Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119463.5 | NP_195035.2 | 1 - 2 | 357 | 74 | |
GenBank mRNA | AK227105.1 | - | 1 - 2 | 254 | 74 | |
AY064961.1 | - | 1 - 2 | 255 | 74 | ||
AY143574.1 | - | 1 - 2 | 230 | 74 | ||
BT005811.1 | - | 1 - 2 | 218 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |