Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADR1-L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036704.2 | NP_001031781.1 | 5 - 6 | 1999 | 73 | |
NM_001342211.1 | NP_001320123.1 | 4 - 5 | 2698 | 73 | ||
GenBank mRNA | AK316970.1 | - | 4 - 5 | 1631 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |