Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203975.1 | NP_001190904.1 | 3 - 4 | 776 | 83 | |
NM_119497.5 | NP_567920.1 | 3 - 4 | 831 | 83 | ||
GenBank mRNA | FJ708762.1 | - | 3 - 4 | 287 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |