Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036707.1 | NP_001031784.1 | 5 - 6 | 534 | 109 | |
NM_001342222.1 | NP_001328574.1 | 4 - 5 | 548 | 106 | ||
NM_119514.4 | NP_567928.1 | 5 - 6 | 640 | 106 | ||
GenBank mRNA | AK221870.1 | - | 5 - 6 | 532 | 106 | |
AY034930.1 | - | 4 - 5 | 395 | 106 | ||
AY088420.1 | - | 4 - 5 | 432 | 106 | ||
AY122986.1 | - | 4 - 5 | 340 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |