Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DRP3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203977.2 | NP_001190906.1 | 12 - 13 | 1809 | 101 | |
NM_119521.5 | NP_567931.1 | 12 - 13 | 1795 | 101 | ||
GenBank mRNA | AB072373.1 | - | 12 - 13 | 1650 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |