Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DRP3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203977.2 | NP_001190906.1 | 6 - 7 | 1202 | 67 | |
NM_119521.5 | NP_567931.1 | 6 - 7 | 1188 | 67 | ||
GenBank mRNA | AB072373.1 | - | 6 - 7 | 1043 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |