Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CER4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342233.1 | NP_001329220.1 | 5 - 6 | 943 | 86 | |
NM_119537.7 | NP_567936.5 | 5 - 6 | 763 | 86 | ||
GenBank mRNA | AY057657.1 | - | 5 - 6 | 749 | 86 | |
AY070065.1 | - | 5 - 6 | 751 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |