Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DML3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125640.1 | NP_001119112.1 | 10 - 11 | 1921 | 109 | |
NM_001342250.1 | NP_001329149.1 | 11 - 12 | 2148 | 109 | ||
NM_001342251.1 | NP_001329150.1 | 8 - 9 | 2173 | 109 | ||
NM_001342252.1 | NP_001329148.1 | 9 - 10 | 2021 | 109 | ||
NM_119567.4 | NP_195132.3 | 10 - 11 | 2075 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |