Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119582.4 | NP_567958.1 | 2 - 3 | 381 | 61 | |
GenBank mRNA | AF133716.1 | - | 2 - 3 | 250 | 61 | |
AY088639.1 | - | 2 - 3 | 271 | 61 | ||
AY090357.1 | - | 2 - 3 | 203 | 61 | ||
BT000842.1 | - | 2 - 3 | 156 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |