Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUC95A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119590.4 | NP_195152.2 | 8 - 9 | 2368 | 66 | |
GenBank mRNA | AY125494.1 | - | 8 - 9 | 2237 | 66 | |
BT002722.1 | - | 8 - 9 | 2233 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |