Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342281.1 | NP_001320134.1 | 3 - 4 | 1268 | 81 | |
NM_119602.2 | NP_567966.1 | 4 - 5 | 1174 | 81 | ||
GenBank mRNA | AY050864.1 | - | 4 - 5 | 1143 | 81 | |
AY117264.1 | - | 4 - 5 | 929 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |