Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342287.1 | NP_001329504.1 | 9 - 10 | 1560 | 59 | |
NM_119614.6 | NP_195175.1 | 9 - 10 | 1519 | 59 | ||
GenBank mRNA | AB014759.1 | - | 9 - 10 | 1396 | 59 | |
BT029161.1 | - | 9 - 10 | 1341 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |