Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COW1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342293.1 | NP_001328041.1 | 4 - 5 | 521 | 77 | |
NM_119624.3 | NP_195184.2 | 3 - 4 | 409 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |