Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT4G34840 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342303.1 | NP_001329682.1 | 6 - 7 | 520 | 96 | |
NM_001342304.1 | NP_001329684.1 | 6 - 7 | 553 | 96 | ||
NM_001342305.1 | NP_001329685.1 | 5 - 6 | 620 | 96 | ||
NM_001342306.1 | NP_001329683.1 | 6 - 7 | 553 | 96 | ||
NM_119650.4 | NP_195210.2 | 6 - 7 | 553 | 96 | ||
GenBank mRNA | AK229730.1 | - | 6 - 7 | 526 | 96 | |
BT010195.1 | - | 6 - 7 | 476 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |