Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334747.1 | NP_001319391.1 | 1 - 2 | 1498 | 138 | |
NM_106341.2 | NP_177816.1 | 2 - 3 | 431 | 138 | ||
NM_179563.3 | NP_849894.1 | 2 - 3 | 843 | 138 | ||
GenBank mRNA | AF487267.1 | - | 1 - 2 | 358 | 138 | |
AY139758.1 | - | 2 - 3 | 529 | 138 | ||
BT003039.1 | - | 1 - 2 | 358 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |