Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335075.1 | NP_001325257.1 | 1 - 2 | 642 | 113 | |
NM_179593.4 | NP_849924.1 | 1 - 2 | 642 | 113 | ||
GenBank mRNA | AY168924.1 | - | 1 - 2 | 535 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |