Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179593.4 | NP_849924.1 | 3 - 4 | 889 | 72 | |
GenBank mRNA | AY168924.1 | - | 3 - 4 | 782 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |