Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179622.4 | NP_849953.2 | 17 - 18 | 2106 | 87 | |
GenBank mRNA | AY337762.1 | - | 17 - 18 | 2089 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |