Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335615.1 | NP_001325303.1 | 3 - 4 | 509 | 116 | |
NM_179653.4 | NP_849984.1 | 3 - 4 | 630 | 116 | ||
GenBank mRNA | AF324344.1 | - | 3 - 4 | 452 | 116 | |
AY063893.1 | - | 3 - 4 | 482 | 116 | ||
AY089963.1 | - | 3 - 4 | 338 | 116 | ||
AY113993.1 | - | 3 - 4 | 338 | 116 | ||
GU198365.1 | - | 3 - 4 | 416 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |