Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HBT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335657.1 | NP_001323708.1 | 2 - 3 | 573 | 81 | |
NM_179663.3 | NP_849994.1 | 2 - 3 | 573 | 81 | ||
GenBank mRNA | AJ487669.1 | - | 2 - 3 | 573 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |