Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLASP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179666.5 | NP_849997.2 | 20 - 21 | 4469 | 105 | |
GenBank mRNA | AY091025.1 | - | 20 - 21 | 4362 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |