Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PFK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179694.4 | NP_850025.1 | 12 - 13 | 1835 | 62 | |
GenBank mRNA | AY070402.1 | - | 12 - 13 | 1503 | 62 | |
AY096578.1 | - | 12 - 13 | 1448 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |