Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336116.1 | NP_001324215.1 | 5 - 6 | 798 | 63 | |
NM_128305.2 | NP_565650.1 | 6 - 7 | 581 | 63 | ||
NM_179770.1 | NP_850101.1 | 6 - 7 | 570 | 63 | ||
GenBank mRNA | AK317300.1 | - | 6 - 7 | 501 | 63 | |
AY072113.1 | - | 6 - 7 | 570 | 63 | ||
AY086056.1 | - | 6 - 7 | 581 | 63 | ||
AY122963.1 | - | 6 - 7 | 549 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |