Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAT12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084501.1 | NP_001077970.1 | 9 - 10 | 2170 | 66 | |
NM_179777.2 | NP_850108.1 | 9 - 10 | 2168 | 66 | ||
GenBank mRNA | AY099545.1 | - | 9 - 10 | 2079 | 66 | |
AY444865.1 | - | 9 - 10 | 1908 | 66 | ||
BT002121.1 | - | 9 - 10 | 1908 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |