Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCRK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036358.2 | NP_001031435.1 | 9 - 10 | 2189 | 86 | |
NM_001336153.1 | NP_001323735.1 | 9 - 10 | 1922 | 86 | ||
NM_001336154.1 | NP_001323736.1 | 9 - 10 | 1663 | 86 | ||
NM_001336155.1 | NP_001323737.1 | 9 - 10 | 2048 | 86 | ||
NM_001336156.1 | NP_001323738.1 | 9 - 10 | 1971 | 86 | ||
NM_179784.2 | NP_850115.1 | 8 - 9 | 2158 | 86 | ||
GenBank mRNA | AY065457.1 | - | 8 - 9 | 1710 | 86 | |
BT004417.1 | - | 8 - 9 | 1540 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |