Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084502.2 | NP_001077971.1 | 10 - 11 | 1623 | 120 | |
NM_001336158.1 | NP_001325347.1 | 9 - 10 | 1509 | 120 | ||
NM_001336159.1 | NP_001325348.1 | 9 - 10 | 1512 | 120 | ||
NM_001336160.1 | NP_001325349.1 | 9 - 10 | 1512 | 120 | ||
NM_179785.3 | NP_850116.1 | 10 - 11 | 1623 | 120 | ||
NM_179786.2 | NP_850117.1 | 10 - 11 | 1623 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Arabidopsis thaliana Probe spans exons |