Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084502.2 | NP_001077971.1 | 8 - 9 | 1072 | 84 | |
NM_001336158.1 | NP_001325347.1 | 7 - 8 | 958 | 84 | ||
NM_001336159.1 | NP_001325348.1 | 7 - 8 | 958 | 84 | ||
NM_001336160.1 | NP_001325349.1 | 7 - 8 | 958 | 84 | ||
NM_179785.3 | NP_850116.1 | 8 - 9 | 1072 | 84 | ||
NM_179786.2 | NP_850117.1 | 8 - 9 | 1072 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |