Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179819.2 | NP_850150.1 | 1 - 2 | 420 | 127 | |
GenBank mRNA | AY081313.1 | - | 1 - 2 | 413 | 127 | |
BT002563.1 | - | 1 - 2 | 108 | 127 | ||
KJ138886.1 | - | 1 - 2 | 108 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |