Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HVT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336303.1 | NP_001325350.1 | 2 - 3 | 1691 | 100 | |
NM_179823.4 | NP_850154.2 | 12 - 13 | 2287 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |