Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APG9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179833.2 | NP_850164.1 | 5 - 6 | 1767 | 90 | |
GenBank mRNA | AB073174.1 | - | 5 - 6 | 1636 | 90 | |
AY075619.1 | - | 5 - 6 | 1615 | 90 | ||
AY099644.1 | - | 5 - 6 | 1615 | 90 | ||
BT000243.1 | - | 4 - 5 | 1303 | 90 | ||
BT000816.1 | - | 4 - 5 | 1303 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |