Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179845.4 | NP_850176.1 | 1 - 2 | 288 | 102 | |
GenBank mRNA | AY065010.1 | - | 1 - 2 | 175 | 102 | |
AY129492.1 | - | 1 - 2 | 123 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |