Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036384.2 | NP_001031461.1 | 5 - 6 | 1140 | 81 | |
NM_001336358.1 | NP_001324731.1 | 5 - 6 | 1166 | 81 | ||
NM_179846.4 | NP_850177.2 | 4 - 5 | 1233 | 81 | ||
GenBank mRNA | AY065265.1 | - | 4 - 5 | 1115 | 81 | |
AY096557.1 | - | 4 - 5 | 1014 | 81 | ||
Z48047.1 | - | 1137 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |