Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036384.2 | NP_001031461.1 | 7 - 8 | 1313 | 151 | |
NM_001202723.1 | NP_001189652.1 | 6 - 7 | 1300 | 151 | ||
NM_001336357.1 | NP_001324733.1 | 5 - 6 | 1580 | 151 | ||
NM_001336358.1 | NP_001324731.1 | 7 - 8 | 1339 | 151 | ||
NM_001336359.1 | NP_001324732.1 | 6 - 7 | 1478 | 151 | ||
NM_179846.4 | NP_850177.2 | 6 - 7 | 1406 | 151 | ||
GenBank mRNA | AY065265.1 | - | 6 - 7 | 1288 | 151 | |
Z48047.1 | - | 1310 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |