Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXPA12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338165.1 | NP_001327548.1 | 1 - 2 | 626 | 69 | |
NM_112405.3 | NP_188156.1 | 2 - 3 | 534 | 69 | ||
GenBank mRNA | BT010937.1 | - | 2 - 3 | 518 | 69 | |
BT011653.1 | - | 2 - 3 | 441 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |