Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SULTR3;4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112469.3 | NP_188220.1 | 3 - 4 | 669 | 101 | |
GenBank mRNA | AB054645.1 | - | 3 - 4 | 669 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |