Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112470.3 | NP_188221.2 | 3 - 4 | 716 | 99 | |
GenBank mRNA | BT008690.1 | - | 3 - 4 | 684 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |