Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCG29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202982.2 | NP_001189911.1 | 2 - 3 | 787 | 67 | |
NM_112505.4 | NP_566543.1 | 2 - 3 | 1033 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |