Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPPalpha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112519.4 | NP_566548.1 | 6 - 7 | 1180 | 74 | |
GenBank mRNA | AY142643.1 | - | 6 - 7 | 984 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |