Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338235.1 | NP_001325855.1 | 1 - 2 | 603 | 91 | |
NM_112521.4 | NP_188271.1 | 1 - 2 | 893 | 91 | ||
GenBank mRNA | AF332394.2 | - | 1 - 2 | 298 | 91 | |
AF332401.1 | - | 1 - 2 | 298 | 91 | ||
AF386948.1 | - | 1 - 2 | 434 | 91 | ||
AY072502.1 | - | 1 - 2 | 298 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |