Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112521.4 | NP_188271.1 | 4 - 5 | 1373 | 78 | |
GenBank mRNA | AF386948.1 | - | 4 - 5 | 914 | 78 | |
AY072502.1 | - | 4 - 5 | 778 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |