Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUM24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112555.2 | NP_566559.1 | 8 - 9 | 1016 | 94 | |
GenBank mRNA | AY045638.1 | - | 8 - 9 | 973 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |