Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112557.4 | NP_188306.2 | 23 - 24 | 3185 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |