Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UPL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125176.1 | NP_001118648.1 | 15 - 16 | 2279 | 146 | |
NM_001202986.1 | NP_001189915.1 | 15 - 16 | 2362 | 146 | ||
NM_112597.4 | NP_188346.2 | 15 - 16 | 2564 | 146 | ||
GenBank mRNA | AY093202.1 | - | 15 - 16 | 2322 | 146 | |
BT010593.1 | - | 15 - 16 | 2087 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |