Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPSN13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035639.2 | NP_001030716.1 | 5 - 6 | 612 | 66 | |
NM_112623.5 | NP_566578.1 | 5 - 6 | 682 | 66 | ||
GenBank mRNA | AY099622.1 | - | 5 - 6 | 550 | 66 | |
BT001249.1 | - | 5 - 6 | 400 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |