Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIPK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112631.3 | NP_566580.1 | 8 - 9 | 1396 | 103 | |
NM_202599.2 | NP_974328.1 | 6 - 7 | 1645 | 103 | ||
GenBank mRNA | BT029495.1 | - | 6 - 7 | 782 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |