Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BSH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202989.1 | NP_001189918.1 | 7 - 8 | 557 | 69 | |
NM_112639.4 | NP_566581.1 | 7 - 8 | 609 | 69 | ||
GenBank mRNA | AK117162.1 | - | 7 - 8 | 551 | 69 | |
BT024620.1 | - | 7 - 8 | 476 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |