Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGD15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338298.1 | NP_001327025.1 | 1 - 2 | 165 | 68 | |
NM_001338299.1 | NP_001327028.1 | 1 - 2 | 165 | 68 | ||
NM_001338300.1 | NP_001327026.1 | 1 - 2 | 165 | 68 | ||
NM_112647.4 | NP_188393.2 | 1 - 2 | 165 | 68 | ||
GenBank mRNA | AK228639.1 | - | 1 - 2 | 165 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |