Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNBT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338307.1 | NP_001326341.1 | 4 - 5 | 1081 | 129 | |
NM_112651.5 | NP_566585.1 | 4 - 5 | 1179 | 129 | ||
GenBank mRNA | AF148541.1 | - | 4 - 5 | 1058 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |