Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TDX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084706.1 | NP_001078175.1 | 6 - 7 | 621 | 75 | |
NM_112669.4 | NP_188415.2 | 6 - 7 | 710 | 75 | ||
GenBank mRNA | AK175494.1 | - | 6 - 7 | 582 | 75 | |
AK227340.1 | - | 6 - 7 | 605 | 75 | ||
AY064251.1 | - | 6 - 7 | 590 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |