Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPK9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338320.1 | NP_001327298.1 | 10 - 11 | 2029 | 91 | |
NM_001338321.1 | NP_001327297.1 | 10 - 11 | 1949 | 91 | ||
NM_112686.4 | NP_566595.1 | 10 - 11 | 1549 | 91 | ||
NM_202602.2 | NP_974331.1 | 9 - 10 | 2072 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |